missing translation for 'onlineSavingsMsg'
Learn More

IRF2BP2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564495
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501564495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501564495 Supplier Bethyl Laboratories Inc Supplier No. A303190A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

IRF2BP2 encodes an interferon regulatory factor-2 (IRF2)binding protein that interacts with the C-terminal transcriptionalrepression domain of IRF2. Alternative splicing results in multipletranscript variants encoding distinct isoforms.

Specifications

Antigen IRF2BP2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene IRF2BP2
Gene Accession No. Q7Z5L9
Gene Alias IRF-2-binding protein 2, interferon regulatory factor 2-binding protein 2
Gene Symbols IRF2BP2
Host Species Rabbit
Immunogen Between 450 and 500
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359948
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less