missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$378.52
Specifications
| Antigen | IRF2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
Description
The recommended shelf life for this product is 1 year from date of receipt.
IRF2 belongs to the interferon regulatory factor family. IRF2 regulates the transcription regulation important to cell proliferation as well as immune responses by binding to the upstream regulatory region of type I IFN and IFNinducible MHC class I genes (the interferon consensus sequence (ICS)) and repression of those genes. However, IRF2 also functions as a transcriptional activator of histone H4 by interactions with NF-kappa B.Specifications
| IRF2 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| DKFZp686F0244, IRF-2 | |
| Between 150 and 200 | |
| Primary | |
| 4° C | |
| IRF2 |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P14316, P23906 | |
| 16363, 3660 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |