missing translation for 'onlineSavingsMsg'
Learn More

Irbesartan Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015429
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015429 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015429 Supplier Merck Emd Millipore Supplier No. PHR195850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 748812-53-5
Linear Formula C25H30N6O2
MDL Number MFCD22380598
Synonym 1-[(1-Oxopentyl)amino]-N-[[2 ′-(2 H-tetrazol-5-yl)[1,1 ′-biphenyl]-4-yl]methyl]cyclopentanecarboxamide
Type Certified Reference Material
Quantity 50 mg
Formula Weight 446.54
Grade Certified Reference Material