missing translation for 'onlineSavingsMsg'
Learn More

Irbesartan Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013026
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013026 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013026 Supplier Merck Emd Millipore Supplier No. PHR14431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Irbesartan is a potent, long-acting 'sartan' class, nonpeptide, angiotensin II receptor antagonist utilized for the management of hypertension as well as heart failure in clinical patients. It shows high selectivity for the AT1 subtype (angiotensin I), and effectively reduces the probabilities of cardiac failure, myocardial infarction, sudden death, etc.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 138402-11-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C25H28N6 O
MDL Number MFCD00864464
Synonym 2-Butyl-3-[[2′-(2 H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4.4 ]non-1-en-4-one
Type Certified Reference Material
Quantity 1 g
Formula Weight 428.53
Grade Certified Reference Material