missing translation for 'onlineSavingsMsg'
Learn More

IR 783 98.0+%, TCI America™

Catalog No. p-7131721
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I1031200MG 200 mg
I10311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I1031200MG Supplier TCI America Supplier No. I1031200MG

Chemical Identifiers

CAS 115970-66-6
Molecular Formula C38H46ClN2NaO6S2
Molecular Weight (g/mol) 749.354
MDL Number MFCD03093208
InChI Key QQIQAVJARACLHE-UHFFFAOYSA-M
Synonym 2-[2-[2-Chloro-3-[2-[1,3-dihydro-3,3-dimethyl-1-(4-sulfobutyl)-2H-indol-2-ylidene]ethylidene]-1-cyclohexen-1-yl]ethenyl]-3,3-dimethyl-1-(4-sulfobutyl)-3H-indolium Inner Salt Sodium Salt, Sodium 4-[2-[2-[2-Chloro-3-[2-[3,3-dimethyl-1-(4-sulfonatobutyl)indo
PubChem CID 71311115
IUPAC Name sodium;4-[(2Z)-2-[(2Z)-2-[2-chloro-3-[(E)-2-[3,3-dimethyl-1-(4-sulfonatobutyl)indol-1-ium-2-yl]ethenyl]cyclohex-2-en-1-ylidene]ethylidene]-3,3-dimethylindol-1-yl]butane-1-sulfonate
SMILES CC1(C2=CC=CC=C2[N+](=C1C=CC3=C(C(=CC=C4C(C5=CC=CC=C5N4CCCCS(=O)(=O)[O-])(C)C)CCC3)Cl)CCCCS(=O)(=O)[O-])C.[Na+]

Specifications

Melting Point 212°C
Quantity 200 mg
Formula Weight 749.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material IR 783