accessibility menu, dialog, popup

UserName

CAS RN 115970-66-6

IUPAC Name: sodium 2-[2-(2-chloro-3-{2-[3,3-dimethyl-1-(4-sulfonatobutyl)-2,3-dihydro-1H-indol-2-ylidene]ethylidene}cyclohex-1-en-1-yl)ethenyl]-3,3-dimethyl-1-(4-sulfonatobutyl)-3H-indol-1-ium
Synonyms: sodium 2-[2-(2-chloro-3-{2-[3,3-dimethyl-1-(4-sulfonatobutyl)indol-2-ylidene]ethylidene}cyclohex-1-en-1-yl)ethenyl]-3,3-dimethyl-1-(4-sulfonatobutyl)indol-1-ium
Molecular Weight (g/mol): 749.35
Molecular Formula: C38H46ClN2NaO6S2
InChi Key: QQIQAVJARACLHE-UHFFFAOYSA-M
SMILES: [Na+].CC1(C)C(=CC=C2CCCC(C=CC3=[N+](CCCCS([O-])(=O)=O)C4=CC=CC=C4C3(C)C)=C2Cl)N(CCCCS([O-])(=O)=O)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results