missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IR 775 Chloride 90.0+%, TCI America™
$301.98 - $896.60
Chemical Identifiers
| CAS | 199444-11-6 |
|---|---|
| Molecular Formula | C32H36Cl2N2 |
| Molecular Weight (g/mol) | 519.55 |
| MDL Number | MFCD03427067,MFCD03427067 |
| InChI Key | BPSIJFMUSNMMAL-UHFFFAOYSA-M |
| Synonym | ir-775 chloride, 2-2-2-chloro-3-2-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-ethylidene-1-cyclohexen-1-yl-ethenyl-1,3,3-trimethyl-3h-indolium chloride, 2-2-2-chloro-3-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene cyclohex-1-en-1-yl ethenyl-1,3,3-trimethyl-3h-indol-1-ium chloride |
| PubChem CID | 71311114 |
| IUPAC Name | 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride |
| SMILES | [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl |
Chemical Identifiers
| 199444-11-6 | |
| 519.55 | |
| BPSIJFMUSNMMAL-UHFFFAOYSA-M | |
| 71311114 | |
| [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl |
| C32H36Cl2N2 | |
| MFCD03427067,MFCD03427067 | |
| ir-775 chloride, 2-2-2-chloro-3-2-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-ethylidene-1-cyclohexen-1-yl-ethenyl-1,3,3-trimethyl-3h-indolium chloride, 2-2-2-chloro-3-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene cyclohex-1-en-1-yl ethenyl-1,3,3-trimethyl-3h-indol-1-ium chloride | |
| 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride |
Specifications
| 199444-11-6 | |
| Green | |
| MFCD03427067,MFCD03427067 | |
| ir-775 chloride, 2-2-2-chloro-3-2-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-ethylidene-1-cyclohexen-1-yl-ethenyl-1,3,3-trimethyl-3h-indolium chloride, 2-2-2-chloro-3-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene cyclohex-1-en-1-yl ethenyl-1,3,3-trimethyl-3h-indol-1-ium chloride | |
| [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl | |
| 519.55 | |
| 519.55 | |
| Crystalline Powder |
| 215°C | |
| C32H36Cl2N2 | |
| 1 g | |
| BPSIJFMUSNMMAL-UHFFFAOYSA-M | |
| 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride | |
| 71311114 | |
| ≥90.0% (N) | |
| IR 775 Chloride |
Safety and Handling
TSCA : No