missing translation for 'onlineSavingsMsg'
Learn More

IR 775 Chloride 90.0+%, TCI America™

Catalog No. I10261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10261G 1 g
I10265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10261G Supplier TCI America Supplier No. I10261G

Chemical Identifiers

CAS 199444-11-6
Molecular Formula C32H36Cl2N2
Molecular Weight (g/mol) 519.55
MDL Number MFCD03427067,MFCD03427067
InChI Key BPSIJFMUSNMMAL-UHFFFAOYSA-M
Synonym ir-775 chloride, 2-2-2-chloro-3-2-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-ethylidene-1-cyclohexen-1-yl-ethenyl-1,3,3-trimethyl-3h-indolium chloride, 2-2-2-chloro-3-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene cyclohex-1-en-1-yl ethenyl-1,3,3-trimethyl-3h-indol-1-ium chloride
PubChem CID 71311114
IUPAC Name 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride
SMILES [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl

Specifications

Melting Point 215°C
Color Green
Quantity 1 g
Formula Weight 519.55
Percent Purity ≥90.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material IR 775 Chloride