missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IQGAP2 Recombinant Rabbit Monoclonal Antibody (23GB4020)
GREENER_CHOICE

Catalog No. MA553672
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553672 100 μL
MA553673 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553672 Supplier Invitrogen™ Supplier No. MA553672

Rabbit Recombinant Monoclonal Antibody

IQGAP2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.

Specifications

Antigen IQGAP2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4020
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IQGAP2
Gene Accession No. Q13576
Gene Alias 4933417J23Rik; A630053O10; AI788777; IQ motif containing GTPase activating protein 2; Iqgap2; Ras GTPase-activating-like protein IQGAP2
Gene Symbols IQGAP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human IQGAP2 (1301-1495AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10788
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less