missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQGAP2 Recombinant Rabbit Monoclonal Antibody (23GB2015)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | IQGAP2 |
|---|---|
| Clone | 23GB2015 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
Description
IQGAP2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.Specifications
| IQGAP2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| IQGAP2 | |
| 4933417J23Rik; A630053O10; AI788777; IQ motif containing GTPase activating protein 2; Iqgap2; Ras GTPase-activating-like protein IQGAP2 | |
| IQGAP2 | |
| Primary | |
| Affinity chromatography |
| 23GB2015 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13576 | |
| 10788 | |
| A synthesized peptide derived from human IQGAP2 (1301-1495AA). | |
| Antibody |