missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IQGAP2 Monoclonal Antibody (K06332_16C7), Invitrogen™

Catalog No. MA555949
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555949 50 μL
MA556329 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555949 Supplier Invitrogen™ Supplier No. MA555949
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: EQTVVTLRNP NAVLTLVDDN LAPEYQKELW DAKKKKEENA RLKNSCISEE ERDAYEELLT QAEIQGNINK VNRQAAVDHI NAVIPEGDPE NTLLALKKPE AQLPAVYPFA AAMYQNELFN LQKQNTMNYL AHEELLIAVE MLSAVALLNQ ALESNDLVSV QNQLRSPAIG LNNLDKAYVE RYANTLLSVK LEVLSQGQDN LSWNEIQNCI DMVNAQIQEE NDRVVAVGYI NEAIDEGNPL RTLETLLLPT ANISDVDPAH AQHYQDVLYH AKSQKLGDSE SVSKVLWLDE IQQAVDDANV

Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.

Specifications

Antigen IQGAP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K06332_16C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene IQGAP2
Gene Accession No. Q13576
Gene Alias 4933417J23Rik; A630053O10; AI788777; IQ motif containing GTPase activating protein 2; Iqgap2; Ras GTPase-activating-like protein IQGAP2
Gene Symbols IQGAP2
Host Species Mouse
Immunogen Recombinant human IQGAP2 (immunogen range is 231-530).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10788
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less