missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IQGAP1 Recombinant Rabbit Monoclonal Antibody (23GB2020)

Description
IQGAP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Spécifications
Spécifications
| Antigène | IQGAP1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB2020 |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gène | IQGAP1 |
| Numéro d’acquisition du gène | P46940, Q9JKF1 |
| Alias du gène | AA682088; Cdc42-Rac1 effector protein; D7Ertd237e; D7Ertd257e; DISP2; DISPB; HUMORFA01; IQ motif containing GTPase activating protein 1; IQGAP1; KIAA0051; mKIAA0051; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs; SAR1 |
| Symboles de gène | IQGAP1 |
| Afficher plus de résultats |