missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ iQ Series™ Men's Lightweight Navy Flame Resistant Ripstop Pant
iQ Series™ Men's Lightweight Navy Flame Resistant Ripstop Pant is stylishly designed to keep you cool and in command all day long. It is NFPA™ 2112 compliant, with a straight-fit, modern jean style.
Supplier: Bulwark™ QP14NV3630
Description
- Sits at the waist, regular in the seat and thigh with a slightly tapered leg
- Jean style cut with one-piece waistband with concealed button closure
- Wide/long belt loops to fit larger belts
- Full crotch gusset and double articulated knees for ease of movement and increased mobility
- Two scoop front pockets and one concealed pocket on right pocket bag, two patch hip pockets, one cargo pocket with snap closures on left leg and one double utility pocket on right leg
- Hazard: Flash Fire, Arc Flash
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 6.9 oz (235 g/m2) Ripstop Twill
- Blend: 65% Cotton/35% Polyester
- Country of Origin: Imported, Made in USA Fabric
Specifications
| 36 x 30 in. | |
| Cotton, Polyester | |
| Stitched | |
| Concealed Button | |
| Category 2 | |
| ASTM F1506, NFPA 2112, NFPA 70E |
| Men's | |
| Navy | |
| 7 | |
| Reusable | |
| 9 |