missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ iQ Series™ Men's Lightweight Navy Flame Resistant Ripstop Pant

Catalog No. p-200000041
Change view
Click to view available options
Size:
28 x 37 in.
30 x 30 in.
30 x 32 in.
30 x 37 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
34 x 37 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
36 x 37 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
38 x 37 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
40 x 37 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
42 x 37 in.
44 x 30 in.
44 x 37 in.
46 x 37 in.
48 x 37 in.
50 x 37 in.
52 x 37 in.
54 x 37 in.
56 x 37 in.
35 product options available for selection
Product selection table with 35 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204335 28 x 37 in.
17204327 30 x 30 in.
17203994 30 x 32 in.
17204194 30 x 37 in.
17103757 32 x 30 in.
17204435 32 x 32 in.
17204427 32 x 34 in.
17204055 34 x 30 in.
17203995 34 x 32 in.
17203999 34 x 34 in.
17204197 34 x 37 in.
17204110 36 x 30 in.
17204202 36 x 32 in.
17103756 36 x 34 in.
17204329 36 x 37 in.
17203996 38 x 30 in.
17204195 38 x 32 in.
17204088 38 x 34 in.
17204199 38 x 37 in.
17204428 40 x 30 in.
17204417 40 x 32 in.
17204418 40 x 34 in.
17204060 40 x 37 in.
17204196 42 x 30 in.
17204003 42 x 32 in.
17204201 42 x 34 in.
17204203 42 x 37 in.
17203990 44 x 30 in.
17204061 44 x 37 in.
17204422 46 x 37 in.
17204198 48 x 37 in.
17204181 50 x 37 in.
17204429 52 x 37 in.
17204432 54 x 37 in.
17204058 56 x 37 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 35 options available.
35 options
Catalog No. 17204335 Supplier Bulwark™ Supplier No. QP14NV 28 37U
Only null left

iQ Series™ Men's Lightweight Navy Flame Resistant Ripstop Pant is stylishly designed to keep you cool and in command all day long. It is NFPA™ 2112 compliant, with a straight-fit, modern jean style.

  • Sits at the waist, regular in the seat and thigh with a slightly tapered leg
  • Jean style cut with one-piece waistband with concealed button closure
  • Wide/long belt loops to fit larger belts
  • Full crotch gusset and double articulated knees for ease of movement and increased mobility
  • Two scoop front pockets and one concealed pocket on right pocket bag, two patch hip pockets, one cargo pocket with snap closures on left leg and one double utility pocket on right leg
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 6.9 oz (235 g/m2) Ripstop Twill
  • Blend: 65% Cotton/35% Polyester
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size 28 x 37 in.
Gender Men's
Material Cotton, Polyester
Color Navy
Seam Type Stitched
No. of Pockets 7
Closure Type Concealed Button
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 9
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less