missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ iQ Series™ Men's Comfort Knit Red Flame Resistant Henley
iQ Series™ Men's Comfort Knit Flame Resistant Henley is CAT 2 AR/FR, NFPA™ 2112 compliant and a casual, comfortable and stylish part of your FR system.
$191.55 - $203.50
Specifications
| Gender | Men's |
|---|---|
| Material | Cotton, Polyester, POD |
| Color | Red |
| Neck Style | Henley |
| Sleeve Style | Raglan Sleeve |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-204-423
|
Bulwark™
QT20RDRGS |
Small |
Each for $203.50
|
|
|||||
|
17-204-083
|
Bulwark™
QT20RDRGM |
Medium |
Each for $203.50
|
|
|||||
|
17-204-330
|
Bulwark™
QT20RDRGL |
Large |
Each for $203.50
|
|
|||||
|
17-204-387
|
Bulwark™
QT20RDRGXL |
X-Large |
Each for $203.50
|
|
|||||
|
17-204-008
|
Bulwark™
QT20RDRGXXL |
2X-Large |
Each for $203.50
|
|
|||||
|
17-204-438
|
Bulwark™
QT20RDRG3XL |
3X-Large |
Each for $203.50
|
|
|||||
|
17-204-012
|
Bulwark™
QT20RDRG4XL |
4X-Large |
Each for $203.50
|
|
|||||
|
17-103-753
|
Bulwark™
QT20RDLNL |
Large (Long) |
Each for $191.55
|
|
|||||
|
17-204-424
|
Bulwark™
QT20RDLNXL |
X-Large (Long) |
Each for $203.50
|
|
|||||
|
17-204-112
|
Bulwark™
QT20RDLNXXL |
2X-Large (Long) |
Each for $203.50
|
|
|||||
|
17-204-059
|
Bulwark™
QT20RDLN3XL |
3X-Large (Long) |
Each for $203.50
|
|
|||||
Description
- With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better
- One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
- Hidden rib knit cuffs to keep sleeves in place
- Moisture-wicking, breathable fabric
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m2)
- Blend: 69% Cotton/25% Polyester/6% Polyoxadiazole
- Closure: Four-button placket
- Product Collection: iQ Series Comfort, iQ Series
- Packaging: Polybag
Specifications
| Men's | |
| Red | |
| Raglan Sleeve | |
| Button | |
| 2 | |
| ASTM F1506, NFPA 2112, NFPA 70E |
| Cotton, Polyester, POD | |
| Henley | |
| Knit | |
| Reusable | |
| Category 2 |