missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ iQ Series™ Men's Comfort Knit Red Flame Resistant Henley

Catalog No. 17204423
Change view
Click to view available options
Size:
2X-Large (Long)
3X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204423 Small
17204083 Medium
17204330 Large
17204387 X-Large
17204008 2X-Large
17204438 3X-Large
17204012 4X-Large
17103753 Large (Long)
17204424 X-Large (Long)
17204112 2X-Large (Long)
17204059 3X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 17204423 Supplier Bulwark™ Supplier No. QT20RD RG S

iQ Series™ Men's Comfort Knit Flame Resistant Henley is CAT 2 AR/FR, NFPA™ 2112 compliant and a casual, comfortable and stylish part of your FR system.

  • With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better
  • One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
  • Hidden rib knit cuffs to keep sleeves in place
  • Moisture-wicking, breathable fabric
  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Home Wash
  • Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m2)
  • Blend: 69% Cotton/25% Polyester/6% Polyoxadiazole
  • Closure: Four-button placket
  • Product Collection: iQ Series Comfort, iQ Series
  • Packaging: Polybag

Specifications

Size Small
Gender Men's
Material Cotton, Polyester, POD
Color Red
Neck Style Henley
Sleeve Style Raglan Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less