missing translation for 'onlineSavingsMsg'
Learn More

Ipriflavone 98.0+%, TCI America™

Catalog No. I06695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06695G 5 g
I066925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06695G Supplier TCI America Supplier No. I06695G

Chemical Identifiers

CAS 35212-22-7
Molecular Formula C18H16O3
Molecular Weight (g/mol) 280.32
MDL Number MFCD00221719
InChI Key SFBODOKJTYAUCM-UHFFFAOYSA-N
Synonym ipriflavone, osten, yambolap, osteofix, 7-isopropoxyisoflavone, iprosten, ipriflavonum, ipriflavona, osteoquine, quinogin
PubChem CID 3747
ChEBI CHEBI:31719
IUPAC Name 3-phenyl-7-(propan-2-yloxy)-4H-chromen-4-one
SMILES CC(C)OC1=CC=C2C(=O)C(=COC2=C1)C1=CC=CC=C1

Specifications

Melting Point 117°C
Color White
Quantity 5 g
Formula Weight 280.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ipriflavone
RTECSNumber DJ3100500
TSCA No