missing translation for 'onlineSavingsMsg'
Learn More

IPA/Hexane, 60:40 v/v, Reagents

Catalog No. 01334931
Change view
Click to view available options
Quantity: 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01334931 Quantity: 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334931 Supplier Labchem Inc Supplier No. CI17110010E6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

IPA/Hexane, 60:40 v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0 , 110-54-3
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Quantity 10 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.7345
Grade Reagent
Chemical Name or Material IPA/Hexane, 60:40 v/v