missing translation for 'onlineSavingsMsg'
Learn More

Iopanoic Acid 98.0+%, TCI America™

Catalog No. p-7131318
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03005G 5 g
I030025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03005G Supplier TCI America Supplier No. I03005G
Only null left

Chemical Identifiers

CAS 96-83-3
Molecular Formula C11H12I3NO2
Molecular Weight (g/mol) 570.935
MDL Number MFCD00038687
InChI Key OIRFJRBSRORBCM-UHFFFAOYSA-N
Synonym 3-Amino-alpha-ethyl-2,4,6-triiodohydrocinnamic Acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropionic Acid, Iodopanoic Acid, 2-(3-Amino-2,4,6-triiodobenzyl)butyric Acid
PubChem CID 3735
ChEBI CHEBI:5951
IUPAC Name 2-[(3-amino-2,4,6-triiodophenyl)methyl]butanoic acid
SMILES CCC(CC1=C(C(=C(C=C1I)I)N)I)C(=O)O

Specifications

Melting Point 153°C
Quantity 5 g
Formula Weight 570.94
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iopanoic Acid
EINECSNumber (9)-0171&(3)-3088
RTECSNumber MW5075000
TSCA No