accessibility menu, dialog, popup

UserName

H3CIIH2NIOHO

CAS RN 96-83-3

IUPAC Name: 2-[(3-amino-2,4,6-triiodophenyl)methyl]butanoic acid
Synonyms: iopanoic acid acide iopanoique colepax acido iopanoico iodopanoic acid bilijodon copanoic acidum iopanoicum acid, iopanoic iopanoic acid (product)
Molecular Weight (g/mol): 570.94
Molecular Formula: C11H12I3NO2
InChi Key: OIRFJRBSRORBCM-UHFFFAOYNA-N
SMILES: CCC(CC1=C(I)C=C(I)C(N)=C1I)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results