missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iopanoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 153°C |
| Quantity | 5 g |
| Formula Weight | 570.94 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Iopanoic Acid |
Safety and Handling
| EINECSNumber | (9)-0171&(3)-3088 |
| RTECSNumber | MW5075000 |
| TSCA | No |