missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ionomycin Calcium Salt, 99%

CAS: 56092-82-1 | C41H70CaO9, C41H70CaO9 | 747.08 g/mol

Supplier:  Thermo Scientific Chemicals J60628LB0

Catalog No. AAJ60628LB0


Only null left

Description

Description

Ionomycin calcium salt is used as a polyether antibiotic and apoptotic degeneration inducer. Ionomycin induces apoptotic degeneration of embryonic cortical neurons and cell cycle arrest at G1 phase. It is more effective than A23187 (sc-3591) and provides a useful tool for increasing intracellular Ca2+ concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5 mg
-30°C to -10°C
99%
C41H70CaO9
Light Sensitive
Ionomycin Calcium Salt, 99%
1416163
[Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O
747.08
White to off-white
Highly selective and potent calcium ionophore
56092-82-1
747.08
Soluble in DMSO (10mg/ml). Also soluble in ethanol or methanol
Crystalline powder
RULWQOBWZOQCOY-SQNLDGQHSA-M
calcium (4R,6S,8S,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,2'R,5S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9,11-dioxodocos-16-en-10-idoate

Chemical Identifiers

56092-82-1
747.08
calcium (4R,6S,8S,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,2'R,5S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9,11-dioxodocos-16-en-10-idoate
C41H70CaO9
RULWQOBWZOQCOY-SQNLDGQHSA-M
[Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only