accessibility menu, dialog, popup

UserName

CaCH3(R)OO(S)CH3(S)CH3OHCO(R)CH3(R)CH3(R)CH3(R)OH(S)CH3(S)HO(S)(S)CH3O(R)(S)CH3O(R)CH3HO(E)

CAS RN 56092-82-1

IUPAC Name: calcium (4R,6S,8S,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,2'R,5S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9,11-dioxodocos-16-en-10-idoate
Synonyms: calcium (4R,6S,8S,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,2'R,5S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9,11-dioxodocos-16-en-10-idoate
Molecular Weight (g/mol): 747.08
Molecular Formula: C41H70CaO9
InChi Key: RULWQOBWZOQCOY-SQNLDGQHSA-M
SMILES: [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O
Color 
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results