missing translation for 'onlineSavingsMsg'
Learn More

Ionic Strength Adjustment Low-Level TISAB II Buffer, SPEX CertiPrep™

Catalog No. ISBUF3500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ISBUF3500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ISBUF3500 Supplier SPEX CertiPrep Group LLC Supplier No. ISBUF3500
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Low Level TISAB II Ionic Strength Adjustment Buffer, 500 mL in H2O, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Ionic Strength Adjustment Buffer
  • Low Level TISAB II
  • 500 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Low Level TISAB II Buffer Solution
Certificate of Analysis With Certificate of Analysis
Concentration 10 M
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Physical Form Liquid
Quantity 500 mL
Solution Type Standard
Packaging Triple-leached LDPE bottle

Product intended for laboratory use only