missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Ion-Exchange Cartridge for Milli-Q™ IQ 7000 Ultrapure Water System

Purification Cartridge for Milli-Q IQ 7000

$1,243.44

Specifications

Flow Rate 2 L/min.
For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System
View More Specs

Products 2
Catalog Number Mfr. No. Type Resin Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Resin Type Price Quantity & Availability  
IPAKQUAA1
View Documents Promotion Details
MilliporeSigma™
IPAKQUAA1
IPAK Quanta™ Polishing Cartridge Ion Exchange Resin, Activated Carbon
Each for $1,243.44
Only null left
 
IPAKMETA1
View Documents Promotion Details
MilliporeSigma™
IPAKMETA1
IPAK Meta™ Polishing Cartridge Ion Exchange Resin
Each for $1,243.44
Only null left
 
Description

Description

Purification Cartridge for Milli-Q IQ 7000

Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments. 
Specifications

Specifications

2 L/min.
Milli-Q IQ 7000 Ultrapure Water System
Videos
SDS
Documents

Documents