missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Ion-Exchange Cartridge for Milli-Q™ IQ 7000 Ultrapure Water System
Description
Purification Cartridge for Milli-Q IQ 7000
Works as a pair with the IPAK Quanta™ polishing cartridge for the Milli-Q™ IQ 7000 system.
Specifications
Specifications
| Flow Rate | 2 L/min. |
| Resin Type | Ion Exchange Resin |
| For Use With (Equipment) | Milli-Q IQ 7000 Ultrapure Water System |
| Type | IPAK Meta™ Polishing Cartridge |