Learn More
Sodium Carbonate/Sodium Bicarbarbonate Eluent Concentrate For Ion Chromatography, 0.18M/0.17M, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier:  SPEX CertiPrep Group LLC ICELCON3100
Catalog No. IC-ELCON3-100
Only null left
Explore available promotions
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Na2CO30.18M
NaHCO30.17M
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Sodium Carbonate/Sodium Bicarbnate Eluent Concentrate
With Certificate of Analysis
Ion Chromatography
Standard Solution
Water
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Certified Reference Material
125 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Product intended for laboratory use only