missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Standard Single Element Ion Standard - Cation, SPEX CertiPrep™

Catalog No. CSMG22Y
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CSMG22Y 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CSMG22Y Supplier SPEX CertiPrep Group LLC Supplier No. CSMG22Y
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Mg1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Magnesium Cation Standard
Packaging Triple-leached LDPE bottle
Certificate of Analysis With Certificate of Analysis
CAS 7439-95-4, 7697-37-2, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.1, 0.2, 99.7
CAS Max % 0.1, 0.2, 99.7
Molecular Formula Mg
Technique Ion Chromatography
MDL Number MFCD00085308
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type Certified Reference Material
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES [Mg]
Solvent or Matrix Nitric Acid
IUPAC Name magnesium
Molecular Weight (g/mol) 24.30
Quantity 125 mL
PubChem CID 5462224
ChEBI CHEBI:25107
Show More Show Less

Product intended for laboratory use only