missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard for Ion Chromatography, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC ASF92X

 View more versions of this product

Catalog No. AS-F9-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
F&superscript_minus;1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Fluoride Standard
With Certificate of Analysis.
0.1,99.9
0.1,99.9
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Certified Reference Material
500 mL
Triple-leached LDPE bottle
7732-18-5,7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Ion Chromatography
Standard Solution
Water
1000μg/mL F-

Chemical Identifiers

7732-18-5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only