missing translation for 'onlineSavingsMsg'
Learn More

Simple Cyanide Reference Standard For Ion Chromatography, H2O/2% KOH, 1000μg/mL:KCN, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC RSCN92Y

 View more versions of this product

Catalog No. RSCN9-2Y


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
CN-1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Cyanide Standard (Simple)
With Certificate of Analysis.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
7732-18-5,7732-18-5
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Certified Reference Material
125 mL
Triple-leached LDPE bottle
0.1,2.0,97.9
0.1,2.0,97.9
Ion Chromatography
Standard Solution
Potassium Hydroxide
For Use In    Matrix    Contents    Cat. No.
EPA Methods 335.2 and 335.3; ASTM Method D2036-19; and Standard Method 4500-CNF.    CN in H2O    1000μg/mL:KCN    RSCN9-2X, -2Y
EPA Method 335.1    CN in H2O    1000μg/mL:K3Fe(CN)6    RSCN9C-2X
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS Documents

Documents

Promotions

Promotions

Product intended for laboratory use only