missing translation for 'onlineSavingsMsg'
Learn More

Iodotrimethylsilane, 95-97%, stabilized

Catalog No. p-149741
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187490050 5 mL Glass bottle
AC187490250 25 mL Glass bottle
AC187491000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC187490050 Supplier Thermo Scientific Chemicals Supplier No. 187490050
Only null left

CAS: 16029-98-4 | C3H9ISi | 200.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 16029-98-4
Molecular Formula C3H9ISi
Molecular Weight (g/mol) 200.1
MDL Number MFCD00001028
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
PubChem CID 85247
IUPAC Name iodo(trimethyl)silane
SMILES C[Si](C)(C)I

Specifications

Color Colorless to Red
Density 1.4000g/mL
Boiling Point 106.0°C
Flash Point −31°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Ex I) (Argentometry)
Packaging Glass bottle
Refractive Index 1.4700 to 1.4800
Linear Formula (CH3)3SiI
Quantity 5 mL
Fieser 08,261; 09,251; 10,216; 11,271; 12,259; 13,329; 16,188; 06,81
Specific Gravity 1.4
Solubility Information Solubility in water: reacts
Stabilizer copper
Formula Weight 200.1
Percent Purity 95 to 97%
Physical Form Liquid
Chemical Name or Material Iodotrimethylsilane, 95-97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-171-0
TSCA TSCA

RUO – Research Use Only