missing translation for 'onlineSavingsMsg'
Learn More

Iodotrimethylplatinum(IV)

Catalog No. AA3928602
Change view
Click to view available options
Quantity:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3928602 0.5 g
AA3928603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3928602 Supplier Thermo Scientific Chemicals Supplier No. 03928602

CAS: 14364-93-3 | C3H9IPt | 367.10 g/mol

They are used as catalysts in organic reactions. It is used in chemical, organic Intermediates research. They are used as platinum group compounds for the preparation of materials suitable for industrial applications, mainly in electronics as coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14364-93-3
Molecular Formula C3H9IPt
Molecular Weight (g/mol) 367.10
MDL Number MFCD00013702
InChI Key ZCSQPOLLUOLHHF-UHFFFAOYSA-M
Synonym iodotrimethylplatinum iv, iodotrimethylplatinum, trimethylplatinum iodide, acmc-1c6hk, trimethylplatinum iv iodide
PubChem CID 11824720
IUPAC Name carbanide;iodoplatinum
SMILES [CH3-].[CH3-].[CH3-].I[Pt]

Specifications

Color Orange to White
Physical Form Crystalline
Quantity 0.5 g
Linear Formula (CH3)3PtI
Solubility Information It is insoluble in water.It is sparingly soluble ether,alcohol,acetone or light petroleum.
Formula Weight 367.09
Odor Odorless
Sensitivity Air & Light Sensitive
Chemical Name or Material Iodotrimethylplatinum(IV)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only