accessibility menu, dialog, popup

UserName

CAS RN 14364-93-3

IUPAC Name: iodoplatinum trimethanide
Synonyms: iodoplatinum trimethanide
Molecular Weight (g/mol): 367.1
Molecular Formula: C3H9IPt
InChi Key: ZCSQPOLLUOLHHF-UHFFFAOYSA-M
SMILES: [CH3-].[CH3-].[CH3-].I[Pt]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results