missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iodonitrotetrazolium violet, 95%

Catalog No. p-7026344
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2230103 1 g
AAB2230106 5 g
AAB2230114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2230103 Supplier Thermo Scientific Chemicals Supplier No. B2230103
Only null left

CAS: 146-68-9 | C19H13ClIN5O2 | 505.70 g/mol

Used for dehydrogenase determination

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 146-68-9
Molecular Formula C19H13ClIN5O2
Molecular Weight (g/mol) 505.70
MDL Number MFCD00011961,MFCD00149999
InChI Key JORABGDXCIBAFL-UHFFFAOYSA-M
Synonym iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chloride
PubChem CID 64957
ChEBI CHEBI:75421
IUPAC Name 2-(4-iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;chloride
SMILES [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1

Specifications

Melting Point ∼231°C (decomposition)
Quantity 1 g
Beilstein 4093224
Sensitivity Light sensitive
Formula Weight 505.70 (Anhydrous)
Percent Purity 95%
Chemical Name or Material Iodonitrotetrazolium violet
Hazard Category H302+H312+H332-H315-H319-H335-H351-H371
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-676-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only