Learn More
Thermo Scientific Chemicals Iodonitrotetrazolium chloride, 98%
CAS: 146-68-9 | C19H13ClIN5O2 | 505.70 g/mol
Supplier: Thermo Scientific Chemicals 122400050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Iodonitrotetrazolium chloride | |
| 231.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00011961,MFCD00149999 | |
| iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chloride | |
| JORABGDXCIBAFL-UHFFFAOYSA-M | |
| 2-(4-iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;chloride | |
| 64957 | |
| 505.7 | |
| Powder, Crystals or Chunks |
| 146-68-9 | |
| White | |
| 97.5% min. (HPLC) | |
| C19H13ClIN5O2 | |
| 5 g | |
| Solubility in water: soluble | |
| [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1 | |
| 505.70 | |
| CHEBI:75421 | |
| 98% |
Chemical Identifiers
| 146-68-9 | |
| 505.70 | |
| JORABGDXCIBAFL-UHFFFAOYSA-M | |
| 64957 | |
| 2-(4-iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;chloride |
| C19H13ClIN5O2 | |
| MFCD00011961,MFCD00149999 | |
| iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chloride | |
| CHEBI:75421 | |
| [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
EINECSNumber : 205-676-2
RUO – Research Use Only