missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iodonitrotetrazolium chloride, 98%

Catalog No. AC122400010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122400010 1 g Glass bottle
AC122400050 5 g Glass bottle
AC122400250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC122400010 Supplier Thermo Scientific Chemicals Supplier No. 122400010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 146-68-9 | C19H13ClIN5O2 | 505.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 146-68-9
Molecular Formula C19H13ClIN5O2
Molecular Weight (g/mol) 505.70
MDL Number MFCD00011961,MFCD00149999
InChI Key JORABGDXCIBAFL-UHFFFAOYSA-M
Synonym iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chloride
PubChem CID 64957
ChEBI CHEBI:75421
IUPAC Name 2-(4-iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;chloride
SMILES [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1

Specifications

Melting Point 231.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 505.7
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Iodonitrotetrazolium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
EINECSNumber 205-676-2

RUO – Research Use Only