missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane, 98%, pure, stabilized with silver

Catalog No. p-149683
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC332121000 100 g Glass bottle
AC332125000 500 g Glass bottle
AC332120025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC332121000 Supplier Thermo Scientific Chemicals Supplier No. 332121000
Only null left

CAS: 75-03-6 | C2H5I | 155.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 75-03-6 , 64-19-7
Molecular Formula C2H5I
Molecular Weight (g/mol) 155.97
MDL Number MFCD00001091
InChI Key HVTICUPFWKNHNG-UHFFFAOYSA-N
Synonym mercury 2+ ion acetic acid
PubChem CID 15337
ChEBI CHEBI:33211
SMILES CCI

Specifications

Melting Point -108.0°C
Color Colorless to Yellow or Pink
Density 1.9500g/mL
Boiling Point 69.0°C to 73.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.5130
Quantity 100 g
Beilstein 01,96
Merck Index 15,3868
Specific Gravity 1.95
Solubility Information Solubility in water: 4g/L (20°C),slowly decomposes in water. Other solubilities: miscible with most other organic solvents,soluble in alcohol,ether,chloroform
IUPAC Name mercury(2+);diacetate
Stabilizer silver
Viscosity 0.592 mPa.s (20°C)
Formula Weight 155.97
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material Iodoethane, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing dif
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
EINECSNumber 200-833-1
RTECSNumber KI4750000
TSCA TSCA
Recommended Storage Product darkens on exposure to light

RUO – Research Use Only