missing translation for 'onlineSavingsMsg'
Learn More

Iodocyclopentane, 97%, stab. with copper

Catalog No. AAL0152206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0152206 5 g
AAL0152214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0152206 Supplier Thermo Scientific Chemicals Supplier No. L0152206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1556-18-9 | C5H9I | 196.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1556-18-9
Molecular Formula C5H9I
Molecular Weight (g/mol) 196.03
MDL Number MFCD00039446
InChI Key PCEBAZIVZVIQEO-UHFFFAOYSA-N
Synonym cyclopentane, iodo, cyclopentyl iodide, jodcyclopentan, cyclopentyliodide, iodo-cyclopentane, iodocyclo-pentane, iodocyclopentane, acmc-1c0qu
PubChem CID 73784
IUPAC Name iodocyclopentane
SMILES IC1CCCC1

Specifications

Density 1.695
Boiling Point 166°C to 167°C
Flash Point 59°C (138°F)
Refractive Index 1.548
Quantity 5 g
UN Number UN1993
Beilstein 1900558
Sensitivity Light sensitive
Formula Weight 196.03
Percent Purity 97%
Chemical Name or Material Iodocyclopentane, stabilized with copper
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 216-311-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only