missing translation for 'onlineSavingsMsg'
Learn More

Iodocyclohexane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. I005610G
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I005610G 10 g
I0056100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I005610G Supplier TCI America Supplier No. I005610G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-62-0
Molecular Formula C6H11I
Molecular Weight (g/mol) 210.06
MDL Number MFCD00003826
InChI Key FUCOMWZKWIEKRK-UHFFFAOYSA-N
Synonym cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i
PubChem CID 12289
IUPAC Name iodocyclohexane
SMILES IC1CCCCC1

Specifications

Boiling Point 180°C
Quantity 10 g
Formula Weight 210.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Iodocyclohexane (stabilized with Copper chip)