accessibility menu, dialog, popup

UserName

I

CAS RN 626-62-0

IUPAC Name: iodocyclohexane
Synonyms: cyclohexane, iodo-
Molecular Weight (g/mol): 210.06
Molecular Formula: C6H11I
InChi Key: FUCOMWZKWIEKRK-UHFFFAOYSA-N
SMILES: IC1CCCCC1
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)