missing translation for 'onlineSavingsMsg'
Learn More

Iodobenzene 99.0+%, TCI America™

Catalog No. I0050500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0050500G 500 g
I005025G 25 g
I0050100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0050500G Supplier TCI America Supplier No. I0050500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 591-50-4
Molecular Formula C6H5I
Molecular Weight (g/mol) 204.01
MDL Number MFCD00001029
InChI Key SNHMUERNLJLMHN-UHFFFAOYSA-N
Synonym benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene
PubChem CID 11575
IUPAC Name iodobenzene
SMILES IC1=CC=CC=C1

Specifications

Melting Point -29°C
Color Yellow
Boiling Point 188°C
Quantity 500 g
Formula Weight 204.01
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Iodobenzene
EINECSNumber (9)-1501
RTECSNumber DA3390000
TSCA Yes