Learn More
Iodobenzene, 98%
CAS: 591-50-4 | C6H5I | 204.01 g/mol
Supplier: Thermo Scientific Chemicals A1278822
Description
Iodobenzene is used to prepare biphenyl and stilbene. It reacts with magnesium to form the Grignard reagent, a phenylmagnesium iodide used in organic synthesis. It finds application as a solid chlorine source by reacting with chlorine to form a complex, iodobenzene dichloride. It serves as a substrate in Sonogashira reaction which is used to form a carbon-carbon bond between a terminal alkyne and an aryl halide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodobenzene is used to prepare biphenyl and stilbene. It reacts with magnesium to form the Grignard reagent, a phenylmagnesium iodide used in organic synthesis. It finds application as a solid chlorine source by reacting with chlorine to form a complex, iodobenzene dichloride. It serves as a substrate in Sonogashira reaction which is used to form a carbon-carbon bond between a terminal alkyne and an aryl halide.
Solubility
Miscible with ethanol, chloroform and ether. Slightly miscible with dimethyl sulfoxide. Immiscible with water.
Notes
Light sensitive. Incompatible with magnesium and strong oxidizing agents.
Specifications
| Iodobenzene | |
| -31°C | |
| 188°C to 189°C | |
| 98% | |
| C6H5I | |
| MFCD00001029 | |
| 1446140 | |
| 14,5029 | |
| Miscible with ethanol,chloroform and ether. Slightly miscible with dimethyl sulfoxide. Immiscible with water. | |
| IC1=CC=CC=C1 | |
| 204.01 | |
| 204.01 |
| 591-50-4 | |
| 1.826 | |
| 74°C (165°F) | |
| Aromatic | |
| 1.62 | |
| 100 g | |
| Light sensitive | |
| benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene | |
| SNHMUERNLJLMHN-UHFFFAOYSA-N | |
| iodobenzene | |
| 11575 | |
| 98% |
Chemical Identifiers
| 591-50-4 | |
| 204.01 | |
| SNHMUERNLJLMHN-UHFFFAOYSA-N | |
| 11575 | |
| IC1=CC=CC=C1 |
| C6H5I | |
| MFCD00001029 | |
| benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene | |
| iodobenzene |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P210-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
H227-H302+H332
EINECSNumber : 209-719-6
RTECSNumber : DA3390000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only