missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetyl chloride, 97%

Catalog No. AAL0442806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0442806 5 g
AAL0442814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0442806 Supplier Thermo Scientific Chemicals Supplier No. L0442806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38020-81-4 | C2H2ClIO | 204.39 g/mol

Iodoacetyl chloride is used for biochemical research purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetyl chloride is used for biochemical research purposes.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents. Reacts with water.

Chemical Identifiers

CAS 38020-81-4
Molecular Formula C2H2ClIO
Molecular Weight (g/mol) 204.39
MDL Number MFCD00013718
InChI Key BSVMPWANOMFSPR-UHFFFAOYSA-N
Synonym iodoacetyl chloride, iodoacetylchloride, acetyl chloride, iodo, jodacetylchlorid, acetylchloride,iodo, iodoacetic acid chloride, acmc-1aewu, iodoacetyl chloride 5g, acetyl chloride, iodo-6ci,9ci
PubChem CID 3084680
IUPAC Name 2-iodoacetyl chloride
SMILES ClC(=O)CI

Specifications

Density 2.28
Boiling Point 48°C to 50°C (12 mmHg)
Quantity 5 g
UN Number UN3265
Beilstein 1739078
Sensitivity Light and moisture sensitive
Solubility Information Miscible with water.
Formula Weight 204.39
Percent Purity 97%
Chemical Name or Material Iodoacetyl chloride
missing translation for 'healthHazard1' H314-H335
missing translation for 'healthHazard2' GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
missing translation for 'healthHazard3' P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 253-748-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only