missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetic Anhydride, 97%

CAS: 54907-61-8 | C4H4I2O3 | 353.88 g/mol

$128.00 - $128.00

Chemical Identifiers

CAS 54907-61-8
Molecular Formula C4H4I2O3
Molecular Weight (g/mol) 353.88
MDL Number MFCD00001080
InChI Key RBNSZWOCWHGHMR-UHFFFAOYSA-N
Synonym iodoacetic anhydride, di iodoacetic anhydride, acmc-20akq5, iodoacetic acid anhydride, 2-iodoacetyl 2-iodoacetate, acetic acid, iodo-, anhydride, acetic acid, 2-iodo-,1,1'-anhydride, iodoacetic anhydride, technical t
PubChem CID 2733254
IUPAC Name (2-iodoacetyl) 2-iodoacetate
SMILES C(C(=O)OC(=O)CI)I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC307592500
SDS View Documents
Thermo Scientific Chemicals
307592500
250 mg
Each for $128.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54907-61-8
353.88
RBNSZWOCWHGHMR-UHFFFAOYSA-N
2733254
C(C(=O)OC(=O)CI)I
C4H4I2O3
MFCD00001080
iodoacetic anhydride, di iodoacetic anhydride, acmc-20akq5, iodoacetic acid anhydride, 2-iodoacetyl 2-iodoacetate, acetic acid, iodo-, anhydride, acetic acid, 2-iodo-,1,1'-anhydride, iodoacetic anhydride, technical t
(2-iodoacetyl) 2-iodoacetate

Specifications

54907-61-8
Yellow
Authentic
Glass bottle
(ICH2CO)2O
250 mg
RBNSZWOCWHGHMR-UHFFFAOYSA-N
(2-iodoacetyl) 2-iodoacetate
2733254
97%
Iodoacetic anhydride
43.0°C to 49.0°C
>110°C
97%
C4H4I2O3
MFCD00001080
iodoacetic anhydride, di iodoacetic anhydride, acmc-20akq5, iodoacetic acid anhydride, 2-iodoacetyl 2-iodoacetate, acetic acid, iodo-, anhydride, acetic acid, 2-iodo-,1,1'-anhydride, iodoacetic anhydride, technical t
C(C(=O)OC(=O)CI)I
353.88
353.88
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only