accessibility menu, dialog, popup

UserName

IOOOI

CAS RN 54907-61-8

IUPAC Name: 2-iodoacetyl 2-iodoacetate
Synonyms: 2-iodoacetyl 2-iodoacetate
Molecular Weight (g/mol): 353.88
Molecular Formula: C4H4I2O3
InChi Key: RBNSZWOCWHGHMR-UHFFFAOYSA-N
SMILES: ICC(=O)OC(=O)CI
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results