missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Iodoacetamide Alkyne

Catalog No. I10189
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10189 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I10189 Supplier Invitrogen™ Supplier No. I10189

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Iodoacetamide Alkyne

Conjugates prepared with the thiol-reactive alkyne, succinimidyl ester can be detected with an azide-containing molecule in a click chemistry reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.

Specifications

Format Solid
Quantity 1 mg
Product Type Iodoacetamide Alkyne
Molecular Weight (g/mol) 223.01 Da
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccated and protect from light.
Chemical Reactivity Thiol
Reactive Group Iodoacetamide
Shipping Condition Room Temperature
Reactive Moiety Alkyl Halide, Iodoacetamide
Label or Dye Alkyne
Label Type Click Chemistry Label
Product Line Molecular Probes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.