missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Iodine-Water-Pyridine-Tetrahydrofuran Oxidizer Solution (0.02 M)

Catalog No. 4347380
Encompass
Change view
Click to view available options
Quantity:
200 mL
450 mL
4 x 180 mL
4 x 450 mL
4 x 2 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4347380 4 x 2 L
4340867 4 x 180 mL
401632 450 mL
4340862 4 x 450 mL
401732 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 4347380 Supplier Applied Biosystems™ Supplier No. 4347380

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains 450 mL 0.02 M Iodine/Water/Pyridine/Tetrahydrofuran Oxidizer Solution.

Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.

Specifications

Quantity 4 x 2 L

For Research Use Only. Not for use in diagnostic procedures.