missing translation for 'onlineSavingsMsg'
Learn More

Iodine Monochloride, Solution, Spectrum™ Chemical

$310.56 - $1605.91

Specifications

Chemical Name or Material Iodine Monochloride
Color Clear, Light Brown
CAS 7790-99-0 , 64-19-7
CAS Max % 1.68,98.32
Grade Chemical Solution
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611066
View Documents
Spectrum Chemical Mfg Cor
I147500MLGL
500 mL Amber Glass Bottle
Each for $310.56
Only null left
 
18611065
View Documents
Spectrum Chemical Mfg Cor
I1474LT
4 L Amber Glass Bottle
Each for $1,605.91
Only null left
 
Description

Description

Specifications

Specifications

Iodine Monochloride
7790-99-0 , 64-19-7
Chemical Solution
To Pass Test
Clear, Light Brown
1.68,98.32
Amber Glass Bottle
Videos
SDS
Documents

Documents

CAUTION: For laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.