missing translation for 'onlineSavingsMsg'
Learn More

Iodine Monochloride, Solution, Spectrum™ Chemical

Catalog No. 18611066
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611066 500 mL Amber Glass Bottle
18611065 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611066 Supplier Spectrum Chemical Mfg Cor Supplier No. I147500MLGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Iodine Monochloride
Color Clear, Light Brown
CAS 7790-99-0, 64-19-7
CAS Max % 1.68, 98.32
Grade Chemical Solution
Packaging Amber Glass Bottle
Suitability To Pass Test
Quantity 500 mL

CAUTION: For laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.