Learn More
Iodine monochloride, 1M soln. in dichloromethane
Description
Iodine monochloride is a useful reagent in organic synthesis and source of I+. It is used as a source of electrophilic iodine in the synthesis of certain aromatic iodides. It also cleaves C-Si bonds. Iodine monochloride will also add to the double bond in alkenes to give chloro-iodo alkanes. It can be dissolved in acetic acid to make Wijs solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Concentration | 1M soln. in dichloromethane |
| Density | 1.42 |
| Linear Formula | ICl |
| Quantity | 500 mL |
| UN Number | UN2922 |
| Sensitivity | Air and light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 162.36 |
| Chemical Name or Material | Iodine monochloride |
Safety and Handling
| Hazard Category | H300-H314-H335-H336-H350 |
| Hazard Statement | GHS H Statement H314-H351 Causes severe skin burns and eye damage. Suspected of causing cancer. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 232-236-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only